ProClin 300 contains two active ingredients: 5-Chloro-2-methyl-4-isothiazolin-3-one (CMIT, CAS RN 26172-55-4) and 2-Methyl-4-isothiazolin-3-one (MIT, CAS RN 2682-20-4).
48919-U
ProClin™ 300
ProClin™ preservative for use in IVD manufacturing, CMIT/MIT 3%, pkg of 110 kg
Synonym(s):
3% solution of 5-chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-4-isothiazolin-3-one
Select a Size
Select a Size
About This Item
Recommended Products
form
liquid
shelf life
3 yr
contains
Alkyl Carboxylate
composition
CMIT/MIT, 3%
packaging
pkg of 110 kg
parameter
operating temperature at <45 °C
matrix
Modified glycol
operating pH range
2.5-8.5
pH range
1.7-3.7
bp
100 °C
mp
-40 °C
solubility
water: soluble at 20 °C
density
1.20 g/cm3
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
244.4 °F - closed cup
Flash Point(C)
118 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What are the active ingredients in ProClin 300?
1 answer-
Helpful?
-
-
What is the concentration of active ingredients in ProClin 300?
1 answer-
ProClin 300 contains a total of 3% active ingredient.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
What is the density of ProClin 300?
1 answer-
The density of ProClin 300 is reported as 8.59 pounds per gallon. The specific gravity at 22°C is reported as 1.03 g/mL.
Helpful?
-
-
What is the matrix of the ProClin 300?
1 answer-
ProClin 300 is a viscous solution containing 93-95 % Modified glycol and 2-3 % Alkyl carboxylate.
Helpful?
-
-
When would I choose to use ProClin 300?
1 answer-
ProClin 300 is best used with production runs of standards, controls, and large volume buffers and solutions. The product is quite viscous and difficult to work with when trying to measure small volumes.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service