Skip to Content
MilliporeSigma
All Photos(1)

Documents

48911-U

Sigma-Aldrich

ProClin 300

ProClin preservative for use in IVD manufacturing, CMIT/MIT 3%, Method Development Kit, pkg of 3 × 5 mL each (three different lots)

Synonym(s):

3% solution of 5-chloro-2-methyl-4-isothiazolin-3-one and 2-Methyl-4-isothiazolin-3-one

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.21

form

liquid

shelf life

3 yr

contains

Alkyl Carboxylate

composition

CMIT/MIT, 3%

packaging

pkg of 3 × 5 mL each (three different lots)

parameter

operating temperature at <45 °C

matrix

Modified glycol

operating pH range

2.5-8.5

pH

4.1(at 100 g/l)

bp

189 °C/1013 hPa

mp

-40 °C

solubility

water: soluble

density

1.030 g/cm3

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

ProClin preservatives are immediately microstatic upon contact with microbial organisms as a result of their ability to quickly penetrate cell membranes and inhibit specific enzymes crucial to cellular respiration. Multiple specific sites of ProClin toxicity prevent microorganisms from developing high levels of resistance, making ProClin highly efficient even over extended storage periods of your IVD components.

Legal Information

ProClin is a trademark of LANXESS Corp.

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada′s Pest Management Regulatory Agency, Turkey′s Biocidal Products Regulation, Korea′s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James P Higham et al.
PloS one, 6(7), e22398-e22398 (2011-07-30)
Nutritional status is a critical element of many aspects of animal ecology, but has proven difficult to measure non-invasively in studies of free-ranging animals. Urinary C-peptide of insulin (UCP), a small polypeptide cleaved in an equimolar ratio from proinsulin when

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service