Data analyzing Antheraxanthin to compare the retention times of 9-cis antheraxanthin and trans antheraxanthin is not available.
47999
9-cis-Antheraxanthin
analytical standard
Synonym(s):
(3S,3′R,5R,6S,9-cis)-5,6-Epoxy-5,6-dihydro-β,β-carotene-3,3′-diol /, (9Z)-Antheraxanthin
Select a Size
Select a Size
About This Item
Recommended Products
grade
analytical standard
Assay
≥95.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
storage temp.
−70°C
SMILES string
CC(/C=C/C1=C(C)C[C@@H](O)CC1(C)C)=C\C=C\C(C)=C\C=C\C=C(C)\C=C\C=C(/C=C/[C@]2(O3)C(C)(C)C[C@H](O)C[C@]23C)C
InChI
1S/C40H56O3/c1-29(17-13-19-31(3)21-22-36-33(5)25-34(41)26-37(36,6)7)15-11-12-16-30(2)18-14-20-32(4)23-24-40-38(8,9)27-35(42)28-39(40,10)43-40/h11-24,34-35,41-42H,25-28H2,1-10H3/b12-11+,17-13+,18-14+,22-21+,24-23+,29-15+,30-16+,31-19+,32-20-/t34-,35+,39-,4
InChI key
OFNSUWBAQRCHAV-CGOFHDSUSA-N
General description
Application
- In petals and pollen extract of Calendula officinalis L. using high-performance liquid chromatography with UV-Vis detection technique.[3]
- In freeze-dried citrus pulp using UV-Vis spectrophotometry and high-performance liquid chromatography-diode array detection-tandem mass spectrometry (HPLC-DAD-MS/MS).[2]
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Do you know what is the difference in retention time for 9-cis antheraxanthin and trans antheraxanthin?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service