Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45815

Supelco

Bromfenvinphos-methyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10BrCl2O4P
CAS Number:
Molecular Weight:
375.97
Beilstein:
6066591
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=O)(OC)O\C(=C\Br)c1ccc(Cl)cc1Cl

InChI

1S/C10H10BrCl2O4P/c1-15-18(14,16-2)17-10(6-11)8-4-3-7(12)5-9(8)13/h3-6H,1-2H3/b10-6+

InChI key

LOOAGTDYKNUDBZ-UXBLZVDNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Konopelko et al.
Medycyna pracy, 40(3), 147-152 (1989-01-01)
The influence of Polfos on the blood pressure in rats was measured by direct and indirect methods. The influence of this pesticide on the heart rate was also examined. Two hours after Polfos administration an increase in blood pressure and
M Konopelko et al.
Medycyna pracy, 39(6), 391-398 (1988-01-01)
The influence of Polfos and its metabolite, 2,4 dichlorophenacyl bromide, on the bleeding time, count of platelets, platelets aggregation, count of erythrocytes, level of haemoglobin and count of leucocytes in rats were studied. The data indicate that Polfos prolonged the
[Sensitivity of ectoparasites of chickens to methylbromfenvinphos (Polwet 5). I. Studies in vitro].
J Złotorzycka et al.
Wiadomosci parazytologiczne, 28(3-4), 387-392 (1982-01-01)
E Lonc et al.
Angewandte Parasitologie, 24(1), 49-55 (1983-02-01)
On the basis of biometrical analysis of various parts of the body in M. gallinae and E. stramineus treated with insecticide (Polwet 5) it was found that only the sizes of abdomen were differently changed in both species. The differentiation
[Sensitivity of ectoparasites of chickens to methylbromfenvinphos (Polwet 5 and Polwet 20). II. Studies in vivo].
J Złotorzycka et al.
Wiadomosci parazytologiczne, 28(3-4), 393-403 (1982-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service