Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

T78603

Sigma-Aldrich

2,3,5-Trimethylphenol

99%

Synonym(s):

Isopseudocumenol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$42.50
100 G
$126.00

$42.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$42.50
100 G
$126.00

About This Item

Linear Formula:
(CH3)3C6H2OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
2042210
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$42.50


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

9.75 mmHg ( 60 °C)

Quality Level

Assay

99%

bp

230-231 °C (lit.)

mp

92-95 °C (lit.)

SMILES string

Cc1cc(C)c(C)c(O)c1

InChI

1S/C9H12O/c1-6-4-7(2)8(3)9(10)5-6/h4-5,10H,1-3H3

InChI key

OGRAOKJKVGDSFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu-Ling Lin et al.
Analytica chimica acta, 871, 57-65 (2015-04-08)
In this study, methacrylic acid (MAA) was incorporated with alkyl methacrylates to increase the hydrophilicity of the synthesized ethylene dimethacrylate-based (EDMA-based) monoliths for separating polar small molecules by capillary LC analysis. Different alkyl methacrylate-MAA ratios were investigated to prepare a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service