Skip to Content
MilliporeSigma

45372

Supelco

Chinomethionate

PESTANAL®, analytical standard

Synonym(s):

Oxythioquinox

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N2OS2
CAS Number:
Molecular Weight:
234.30
Beilstein:
526833
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cc1ccc2nc3SC(=O)Sc3nc2c1

InChI

1S/C10H6N2OS2/c1-5-2-3-6-7(4-5)12-9-8(11-6)14-10(13)15-9/h2-4H,1H3

InChI key

FBQQHUGEACOBDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 2 - Skin Sens. 1A - STOT RE 2 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Carrera et al.
Nutrition and metabolism, 24(2), 76-90 (1980-01-01)
The effects of oxythioquinox (200 mg/kg fresh food for 30 days) on the growth, feeding, body composition and weight of various organs were studied in rats receiving isocaloric diets containing varying amounts of alimentary lipid (3, 18, 35 and 49%).
K Sasaki et al.
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan, 42(4), 273-277 (2002-01-31)
Method-performance studies were conducted for the notified revised analytical method of chinomethionat in agricultural products by interlaboratory study. Six laboratories analyzed unpolished rice, cabbage, squash, lettuce and orange spiked with 0.1 microgram/g of chinomethionat in replicate. Mean values of recovery
G Carrera et al.
Toxicology letters, 19(1-2), 159-164 (1983-10-01)
Oxythioquinox, administered in solution in the dietary lipids at 400 mg/kg fresh food for 21 days, caused liver hypertrophy with severe steatosis. The levels of liver triglycerides increased 40-fold, phospholipids 2-fold and cholesterol 7-fold respectively. The steatosis was thought to
G Carrera et al.
Comptes rendus des seances de l'Academie des sciences. Serie III, Sciences de la vie, 296(21), 1001-1004 (1983-01-01)
The amount of nitrogen excreted in urines is higher in pair-fed Rats subjected to a diet containing 400 mg/kg of oxythioquinox. This is a consequence of specific effect of oxythioquinox on protein catabolism and not to a renal failure as
R T Krause et al.
Journal - Association of Official Analytical Chemists, 66(4), 1018-1022 (1983-07-01)
The AOAC multiresidue method for nonpolar pesticide residues in nonfatty foods has been coupled with a high performance liquid chromatographic (HPLC)-fluorometric system for determining quinomethionate residues on apples and oranges. Quinomethionate is extracted with acetonitrile, and coextractives are removed with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service