Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43344

Supelco

Nordihydrocapsiate

analytical standard

Synonym(s):

(4-Hydroxy-3-methoxyphenyl)methyl 7-methyloctanoate, 4-Hydroxy-3-methoxybenzyl 7-methyloctanoate, NDCT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26O4
CAS Number:
Molecular Weight:
294.39
Beilstein:
8272135
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage condition

under inert gas

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.495-1.505

format

neat

storage temp.

−20°C

SMILES string

OC1=C(OC)C=C(COC(CCCCCC(C)C)=O)C=C1

InChI

1S/C17H26O4/c1-13(2)7-5-4-6-8-17(19)21-12-14-9-10-15(18)16(11-14)20-3/h9-11,13,18H,4-8,12H2,1-3H3

InChI key

BXBVPYSHEOQGHP-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

No-Joon Song et al.
Experimental & molecular medicine, 49(7), e353-e353 (2017-07-08)
Adipose tissue is a central metabolic organ that controls energy homeostasis of the whole body. White adipose tissue (WAT) stores excess energy in the form of triglycerides, whereas brown adipose tissue (BAT) dissipates energy in the form of heat through
Jose M Rubio et al.
The lancet. Psychiatry, 7(9), 749-761 (2020-08-24)
Most individuals with schizophrenia-spectrum disorders have relapses, which increase the risk of morbidity and mortality. Because non-adherence to antipsychotic maintenance treatment could affect more than half of individuals with schizophrenia-spectrum disorders, psychosis relapse can often be confounded by unnoticed treatment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service