Skip to Content
MilliporeSigma
All Photos(1)

Documents

05088

Supelco

Dihydrocapsiate

analytical standard

Synonym(s):

(4-Hydroxy-3-methoxyphenyl)methyl 8-methylnonanoate, 4-Hydroxy-3-methoxybenzyl 8-methylnonanoate, DCT, Vanillyl 8-methylnonanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H28O4
CAS Number:
Molecular Weight:
308.41
Beilstein:
7931830
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage condition

under inert gas

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

OC1=C(OC)C=C(COC(CCCCCCC(C)C)=O)C=C1

InChI

1S/C18H28O4/c1-14(2)8-6-4-5-7-9-18(20)22-13-15-10-11-16(19)17(12-15)21-3/h10-12,14,19H,4-9,13H2,1-3H3

InChI key

RBCYRZPENADQGZ-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service