Skip to Content
MilliporeSigma
All Photos(1)

Documents

41624

Sigma-Aldrich

Dimethyl sulfide

puriss., ≥99.0% (GC)

Synonym(s):

DMS, Methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S
CAS Number:
Molecular Weight:
62.13
Beilstein:
1696847
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.1 (vs air)

vapor pressure

26.24 psi ( 55 °C)
7.79 psi ( 20 °C)

grade

puriss.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

402 °F

expl. lim.

19.7 %

refractive index

n20/D 1.435 (lit.)
n20/D 1.435

bp

38 °C (lit.)

mp

−98 °C (lit.)

solubility

H2O: soluble 7.28 g/L at 20 °C

density

0.846 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CSC

InChI

1S/C2H6S/c1-3-2/h1-2H3

InChI key

QMMFVYPAHWMCMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-32.8 °F - closed cup

Flash Point(C)

-36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nguyen BC, et al.
Marine Chemistry, 133-141 (1988)
B P Lomans et al.
Applied and environmental microbiology, 65(8), 3641-3650 (1999-07-31)
A newly isolated methanogen, strain DMS1(T), is the first obligately anaerobic archaeon which was directly enriched and isolated from a freshwater sediment in defined minimal medium containing dimethyl sulfide (DMS) as the sole carbon and energy source. The use of
Sheree Yau et al.
The ISME journal, 7(10), 1944-1961 (2013-04-27)
Organic Lake is a shallow, marine-derived hypersaline lake in the Vestfold Hills, Antarctica that has the highest reported concentration of dimethylsulfide (DMS) in a natural body of water. To determine the composition and functional potential of the microbial community and
Hayley E Arnold et al.
Global change biology, 19(4), 1007-1016 (2013-03-19)
The production of the marine trace gas dimethyl sulfide (DMS) provides 90% of the marine biogenic sulfur in the atmosphere where it affects cloud formation and climate. The effects of increasing anthropogenic CO2 and the resulting warming and ocean acidification
K S Carslaw et al.
Nature, 503(7474), 67-71 (2013-11-10)
The effect of anthropogenic aerosols on cloud droplet concentrations and radiative properties is the source of one of the largest uncertainties in the radiative forcing of climate over the industrial period. This uncertainty affects our ability to estimate how sensitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service