Skip to Content
MilliporeSigma
Dimethyl sulfide analytical standard

Dimethyl sulfide

Dimethyl sulfide analytical standard
Synonym(s):
DMS, Methyl sulfide, Dimethyl sulfide
Linear Formula:
(CH3)2S
CAS Number:
Molecular Weight:
62.13
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique