Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41474

Supelco

Levulinic acid

analytical standard

Synonym(s):

4-Oxopentanoic acid, 4-Oxovaleric acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$94.20

$94.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$94.20

About This Item

Linear Formula:
CH3COCH2CH2COOH
CAS Number:
Molecular Weight:
116.12
Beilstein:
506796
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$94.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor pressure

1 mmHg ( 102 °C)

Assay

≥97.5% (GC)
97.5-102.5% (T)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

245-246 °C (lit.)

mp

30-33 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)CCC(O)=O

InChI

1S/C5H8O3/c1-4(6)2-3-5(7)8/h2-3H2,1H3,(H,7,8)

InChI key

JOOXCMJARBKPKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Levulinic acid (LA), is a biogenic product of hexose acid hydrolysis, produced from renewable or bio-based resources.[1][2] LA is reported to react with a variety of compounds to result in products, which are widely used in pharmaceuticals, personal care products, electronics, plasticizers, antifouling compounds, fuels, solvents, polymers, anti-freeze agents, batteries, coatings, biologically active materials, corrosion inhibitors, adsorbents, etc.[2] LA is also employed as a chemical intermediate in the production of biodegradable polymers, fuel extenders, herbicides, flavors, antibiotics and useful 5-carbon compounds.[1]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Levulinic acid may be used as an analytical reference standard
for the quantification of the analyte in the following:
  • Soy sauce using liquid chromatography coupled to mass spectrometry (LC–MS).[3]
  • Liquid food samples using gas chromatography with flame ionization detection (GC-FID).[4]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Levulinic acid
Hachula B, et al.
Acta Crystallographica Section E, Structure Reports Online, 69(9) (2013)
A Review on the conversion of levulinic acid and its esters to various useful chemicals
Adeleye TA, et al.
AIChE Journal, 7(2), 165-165 (2019)
Determination of levulinic acid in soy sauce by liquid chromatography with mass spectrometric detection
Sano A, et al.
Food Chemistry, 105(3), 1242-1247 (2007)
A rapid gas chromatographic method for direct determination of short-chain (C2?C12. volatile organic acids in foods
Yang H-M and Choong M-Y
Food Chemistry, 75(1), 101-108 (2001)
F Monlau et al.
Bioresource technology, 187, 379-386 (2015-04-13)
The robust supramolecular structure of biomass often requires severe pretreatments conditions to produce soluble sugars. Nonetheless, these processes generate some inhibitory compounds (i.e. furans compounds and aliphatic acids) deriving mainly from sugars degradation. To avoid the inhibition of the biological

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service