Skip to Content
MilliporeSigma
Levulinic acid analytical standard

Levulinic acid

Levulinic acid analytical standard
Synonym(s):
4-Oxopentanoic acid, 4-Oxovaleric acid, Levulinic acid
Linear Formula:
CH3COCH2CH2COOH
CAS Number:
Molecular Weight:
116.12
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing