Skip to Content
MilliporeSigma
All Photos(1)

Documents

38483

Sigma-Aldrich

1,2-Didodecanoyl-sn-glycero-3-phosphocholine

≥99.0% (TLC)

Synonym(s):

(7R)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxododecyl)oxy]-3,5,9-trioxa-4-phosphaheneicosan-1-aminium 4-oxide, inner salt, 1,2-Dilauroyl-sn-glycero-3-phosphocholine, 1,2-Dilauroyl-sn-glycero-3-phosphorylcholine, 3-sn-Phosphatidylcholine, 1,2-dilauroyl, L-β,γ-Dilauroyl-α-lecithin, DLPC, PC(12:0/12:0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C32H64NO8P
CAS Number:
Molecular Weight:
621.83
Beilstein:
5676925
EC Number:
MDL number:
UNSPSC Code:
12352211

Assay

≥99.0% (TLC)

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O)=O

InChI

1S/C32H64NO8P/c1-6-8-10-12-14-16-18-20-22-24-31(34)38-28-30(29-40-42(36,37)39-27-26-33(3,4)5)41-32(35)25-23-21-19-17-15-13-11-9-7-2/h30H,6-29H2,1-5H3/t30-/m1/s1

InChI key

IJFVSSZAOYLHEE-SSEXGKCCSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eman Abd et al.
Skin pharmacology and physiology, 29(1), 24-30 (2015-11-06)
A range of vesicles is now widely used to carry various solutes into and through the epidermis. These usually have the active solute encapsulated within and may be modified to confer flexibility and skin penetration enhancement. Here, we compared the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service