Skip to Content
MilliporeSigma
1,2-Didodecanoyl-sn-glycero-3-phosphocholine ≥99%, synthetic

1,2-Didodecanoyl-sn-glycero-3-phosphocholine

1,2-Didodecanoyl-sn-glycero-3-phosphocholine ≥99%, synthetic
Synonym(s):
(7R)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[(1-oxododecyl)oxy]-3,5,9-trioxa-4-phosphaheneicosan-1-aminium 4-oxide, inner salt, 1,2-Dilauroyl-sn-glycero-3-phosphocholine, 1,2-Dilauroyl-sn-glycero-3-phosphorylcholine, 3-sn-Phosphatidylcholine, 1,2-dilauroyl, L-β,γ-Dilauroyl-α-lecithin, DLPC, PC(12:0/12:0)
Linear Formula:
C32H64NO8P
CAS Number:
Molecular Weight:
621.83
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing