Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37214

Supelco

Peroxide 25 Test Sticks Quantofix®

0.5-25 mg/L (H2O2)

Synonym(s):

Quantofix® Peroxide 25 Test sticks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
Pricing and availability is not currently available.

measuring range

0.5-25 mg/L (H2O2)

application(s)

environmental

General description

Contents:
Tube with 100 test sticks.

Storage:
Avoid exposing the strips to sunlight and moisture. Keep container cool and dry (store below +30 °C).

Gradation:
0 - 0.5 - 2 - 5 - 10 - 25 mg/l H2O2

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service