Skip to Content
MilliporeSigma
All Photos(2)

Documents

Z683108

Sigma-Aldrich

XploSens PS peroxides detection strips

quickly detects peroxides in organic and aqueous solutions

Synonym(s):

peroxide indicator strips, simple peroxide detection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034
NACRES:
NB.35

description

semi-quantitative

specific analyte(s)

hydrogen peroxide

reaction suitability

reaction type: Redox Reactions

packaging

pack of 50 test strips

manufacturer/tradename

XploSafe 3003

detection method

colorimetric

General description

XploSens peroxide detection strips quickly detects peroxides in organic and aqueous solutions. 50 test strips.

XploSens features:
  • One step trace detection of peroxides in organic and aqueous solutions
  • Simple and effective - rapid and distinct color change
  • Room temperature storage
  • Gradations: 0-50, 50-100, 100-250, 250-350, 350-500 ppm

Directions: Immerse the pad of the test strip into the solution for 5 seconds and remove. An appearance of a yellow color on the pad indicates that trace amounts of peroxides are present. The development of the yellow color may take up to 3 minutes and varies with the solvent being tested.

Legal Information

XploSens PS is a trademark of XploSafe, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service