Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

37066

Supelco

Calcium test kit Quantofix®

10-100 mg/L (Ca2+)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319
NACRES:
NA.21

description

semi-quantitative

specific analyte(s)

calcium

measuring range

10-100 mg/L (Ca2+)

application(s)

agriculture
environmental
food and beverages
general analytical

detection method

colorimetric

General description

Contents:
Kit including reagents and test sticks for 60 tests.

Storage:
Avoid exposing the sticks to sunlight and moisture. Store the kit below +30 °C in a dry place.

Gradation:
0 − 10 − 25 − 50 − 100 mg/L Ca2+

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service