Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

32957

Supelco

Cresyl diphenyl phosphate

PESTANAL®, analytical standard

Synonym(s):

Diphenyl cresyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17O4P
CAS Number:
Molecular Weight:
340.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

description

mixture of isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cccc(OP(=O)(Oc2ccccc2)Oc3ccccc3)c1

InChI

1S/C19H17O4P/c1-16-9-8-14-19(15-16)23-24(20,21-17-10-4-2-5-11-17)22-18-12-6-3-7-13-18/h2-15H,1H3

InChI key

FJKVRVFGUXMMAO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

464.0 °F - open cup

Flash Point(C)

240 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Vainiotalo et al.
Toxicology, 44(1), 31-44 (1987-04-01)
A commercial cresyl diphenyl phosphate preparation was analyzed to contain approximately 35% of triphenyl phosphate, 45% of cresyl diphenyl phosphates, 18% of dicresyl phenyl phosphates and 2% of tricresyl phosphates. The product was almost free of the o-cresyl isomers as
D C Muir et al.
Journal - Association of Official Analytical Chemists, 64(1), 79-84 (1981-01-01)
Several techniques were evaluated for extracting triphenyl phosphate (TPP), 14C-labeled TPP, cresyl diphenyl phosphate, and tricresyl phosphate isomers (o-TCP, m-TCP, and p-TCP) from fish and sediment samples. Extracts of fish samples were cleaned up by gel permeation chromatography/alumina column chromatography;
M Lotti et al.
Archives of toxicology, 45(4), 263-271 (1980-10-01)
The effects of small repeated oral doses of mono-2-cresyl diphenyl phosphate (MOCP, 2.5 mg/kg/day) on hen brain and spinal cord neurotoxic esterase (NTE) were measured. The enzyme levels were depressed to about 40% and 55% of normal respectively and maintained
B Sala et al.
Environmental research, 172, 289-295 (2019-03-02)
This study evaluates for the first time organophosphorus flame retardant (OPFR) occurrence in the Alboran Sea delphinids (Spain). OPFRs were detected in all the individuals with concentration levels up to 24.7 µg/g lw. Twelve out of sixteen tested analytes were detected
Fengqiong Shi et al.
Environment international, 139, 105653-105653 (2020-05-04)
Numerous studies have documented that the general population is widely exposed to organophosphate esters (OPEs), yet studies on the emissions of OPEs in the industrial application processes and their occupational exposure are scarce. The aim of this study was to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service