MilliporeSigma
  • Acute biological effects of commercial cresyl diphenyl phosphate in rats.
Acute biological effects of commercial cresyl diphenyl phosphate in rats.
Toxicology (1987-04-01)
S Vainiotalo, E Verkkala, H Savolainen, J Nickels, A Zitting
ABSTRACT
A commercial cresyl diphenyl phosphate preparation was analyzed to contain approximately 35% of triphenyl phosphate, 45% of cresyl diphenyl phosphates, 18% of dicresyl phenyl phosphates and 2% of tricresyl phosphates. The product was almost free of the o-cresyl isomers as revealed by the analysis of its alkaline hydrolysis products. A single intraperitoneal injection (150 or 300 mg/kg) caused an induction of microsomal cytochrome P-450 in the liver of Wistar rats with a concomitant increase in the activities of mixed function monooxygenases and proliferation of smooth endoplasmic reticulum 24 h after the treatment. These effects were not detected in the kidneys. The morphological changes in hepatocytes included the enlargement of nuclei and mitochondria with increased cristae. The hepatic morphology returned to normal 2 weeks after the treatment. The activity of pseudocholine esterase in blood was inhibited 4 h and 24 h after the injection but the effect levelled off. The concentration of the organophosphates in blood and liver decreased rapidly with only traces detected in blood after 24 h. No effects on the activities of cerebral and muscle acetylcholine esterase were observed. The treatment (300 mg/kg) inhibited the brain--2',3'-cyclic nucleotide 3'-phosphohydrolase through the 2-week observation period associated with demyelination in peripheral nerves.