Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

32854

Supelco

Azaperone-d4

VETRANAL®, analytical standard

Synonym(s):

1-(4-Fluorophenyl-d4)-4-(4-(2-pyridinyl)-1-piperazinyl)-1-butanone, 4′-Fluoro-4-[4-(2-pyridyl)-1-piperazinyl]butyrophenone-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19D4H18FN3O
CAS Number:
Molecular Weight:
331.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+4

SMILES string

[2H]c1c([2H])c(c([2H])c([2H])c1F)C(=O)CCCN2CCN(CC2)c3ccccn3

InChI

1S/C19H22FN3O/c20-17-8-6-16(7-9-17)18(24)4-3-11-22-12-14-23(15-13-22)19-5-1-2-10-21-19/h1-2,5-10H,3-4,11-15H2/i6D,7D,8D,9D

InChI key

XTKDAFGWCDAMPY-YKVCKAMESA-N

General description

Azaperone-d4 is an isotopically labeled compound of tranquilizer drug, azaperone.[1]

Application

Azaperone-d4 may be used as an internal standard for the determination of azaperone and azaperol in animal muscles by liquid chromatography-tandem mass spectrometry (LC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robustness testing in the determination of seven drugs in animal muscle by liquid chromatography-tandem mass spectrometry.
Oca Casado M, et al.
Chemometrics and Intelligent Laboratory Systems (2016)
Salomé Martínez-Morcillo et al.
International journal of hygiene and environmental health, 223(1), 10-21 (2019-11-11)
This study assessed the presence of 27 pharmaceutically active compounds belonging to common therapeutic groups (cardiovascular, antiashmatic, psychoactive, diuretic, analgesic/anti-inflammatory, and antibiotic drugs) in the tissues of representative seafood species of bivalves, cephalopods, arthropods, and fish of high economic importance
Lenise Guimarães de Oliveira et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(1), 32-39 (2016-11-22)
A method for the determination of five sedatives and 14 β-blocker residues in swine, bovine and equine kidney was validated. Samples were extracted with acetonitrile and purified using dispersive solid phase extraction (d-SPE) with Celite 545 with subsequent analysis by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service