Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

211280

Sigma-Aldrich

Capsazepine

A competitive antagonist of Capsaicin and resiniferatoxin. Capsazepine (1 µM) blocks capsaicin (60 nM) stimulated firing of C-fibers.

Synonym(s):

Capsazepine, N-[2-(4-Chlorophenyl)ethyl]-1,3,4,5-tetrahydro-7,8-dihydroxy-2H-2-benzazepine-2-carbothioamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$491.00

$491.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$491.00

About This Item

Empirical Formula (Hill Notation):
C19H21ClN2O2S
CAS Number:
Molecular Weight:
376.90
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$491.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: soluble
methanol: soluble

shipped in

ambient

storage temp.

10-30°C

SMILES string

S=C(N2CCCc3c(cc(c(c3)O)O)C2)NCCc1ccc(cc1)Cl

InChI

1S/C19H21ClN2O2S/c20-16-5-3-13(4-6-16)7-8-21-19(25)22-9-1-2-14-10-17(23)18(24)11-15(14)12-22/h3-6,10-11,23-24H,1-2,7-9,12H2,(H,21,25)

InChI key

DRCMAZOSEIMCHM-UHFFFAOYSA-N

General description

A competitive antagonist of Capsaicin (Cat. No. 211274) and resiniferatoxin. Capsazepine (1 µM) blocks capsaicin (60 nM) stimulated firing of C-fibers.
A competitive antagonist of the sensory neuron excitants capsaicin (Cat. No. 211274) and resiniferatoxin (Cat. No. 553975). Capsazepine (1 µM) blocks capsaicin (60 nM) stimulated firing of C-fibers.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A competitive antagonist of Capsaicin and resiniferatoxin
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Fox, A.J., et al. 1995. Neuroscience 67, 741.
Ellis, J.L., and Undem, B.J. 1994. J. Pharmacol. Exp. Ther. 268, 85.
Walpole, C.S.J., et al. 1994. J. Med. Chem.37, 1942.
Szallasi, A., et al. 1993. J. Pharmacol. Exp. Therap.267, 728.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service