Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23514

Sigma-Aldrich

2-Chloro-1,3,2-benzodioxaphosphorin-4-one

purum, ≥95.0% (AT)

Synonym(s):

Salicyl chlorophosphite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4ClO3P
CAS Number:
Molecular Weight:
202.53
Beilstein:
1311951
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

grade

purum

Assay

≥95.0% (AT)

bp

127-128 °C/11 mmHg (lit.)

mp

36-40 °C (lit.)

storage temp.

2-8°C

SMILES string

ClP1OC(=O)c2ccccc2O1

InChI

1S/C7H4ClO3P/c8-12-10-6-4-2-1-3-5(6)7(9)11-12/h1-4H

InChI key

BVOITXUNGDUXRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Phosphitylating agent for nucleosides and carbohydrates; Synthesis of triphosphates and 1-thiotriphosphates from alcohols

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.P.G. Hermans et al.
Rec. Trav. Chim., 105, 510-510 (1986)
Yuka Kato et al.
Nucleic acids research, 33(9), 2942-2951 (2005-05-26)
The synthesis of the triphosphates of 4'-thiouridine and 4'-thiocytidine, 4'-thioUTP (7; thioUTP) and 4'-thioCTP (8; thioCTP), and their utility for SELEX (systematic evolution of ligands by exponential enrichment) is described. The new nucleoside triphosphate (NTP) analogs 7 and 8 were
J.E. Marugg et al.
Tetrahedron Letters, 27, 2661-2661 (1986)
P. Westerduin et al.
Tetrahedron Letters, 27, 6271-6271 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service