Skip to Content
MilliporeSigma
2-Chloro-1,3,2-benzodioxaphosphorin-4-one 95%

2-Chloro-1,3,2-benzodioxaphosphorin-4-one

2-Chloro-1,3,2-benzodioxaphosphorin-4-one 95%
Synonym(s):
Salicyl chlorophosphite
Empirical Formula (Hill Notation):
C7H4ClO3P
CAS Number:
Molecular Weight:
202.53

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing