Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

222313

Sigma-Aldrich

Calcium chloride

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCl2
CAS Number:
Molecular Weight:
110.98
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

technical grade

vapor pressure

0.01 mmHg ( 20 °C)

mp

772 °C (lit.)

SMILES string

[Cl-].[Cl-].[Ca++]

InChI

1S/Ca.2ClH/h;2*1H/q+2;;/p-2

InChI key

UXVMQQNJUSDDNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell specific aptamer-photosensitizer conjugates as a molecular tool in photodynamic therapy.
Prabodhika Mallikaratchy et al.
ChemMedChem, 3(3), 425-428 (2007-12-07)
Patricia W Kimani et al.
Respiratory research, 10, 119-119 (2009-11-27)
Platelet-derived growth factor A (PDGF-A) signals solely through PDGF-Ralpha, and is required for fibroblast proliferation and transdifferentiation (fibroblast to myofibroblast conversion) during alveolar development, because pdgfa-null mice lack both myofibroblasts and alveoli. However, these PDGF-A-mediated mechanisms remain incompletely defined. At
P Ranjit et al.
Vascular pharmacology, 71, 102-107 (2015-04-15)
Research and routine laboratory assessment of clot integrity can be time consuming, expensive, and cannot be batched as it is generally performed in real time. To address these issues, we developed and validated a micro-titre based assay to quantify thrombogenesis
Hadi Shafiee et al.
Scientific reports, 5, 9919-9919 (2015-06-06)
We report a biosensing platform for viral load measurement through electrical sensing of viruses on a flexible plastic microchip with printed electrodes. Point-of-care (POC) viral load measurement is of paramount importance with significant impact on a broad range of applications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service