Skip to Content
MilliporeSigma
All Photos(1)

Documents

20886

Hamilton® HPLC syringes

1702RNR, volume 25 μL, needle size 22s ga (blunt tip), needle L 51 mm (2 in.)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

blunt tip stainless steel needle (point style 3)
glass body

needle size

22s ga (blunt tip)

feature

blunt tip

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 80265

needle L

51 mm (2 in.)

volume

25 μL

compatibility

for use with Rheodyne
for use with SSI
for use with Valco VISF-2

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service