Skip to Content
MilliporeSigma
All Photos(1)

Documents

18405

Sigma-Aldrich

Vacuum grease

Synonym(s):

Grease for vacuum lines, Stopcock grease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

form

paste

Application

Vacuum grease has been used to create a water-tight seal of the coverslips during fluorescence resonance energy transfer (FRET) imaging. It has also been used to seal the input and output channels during optical torque wrench experiments.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Tang et al.
ACS nano, 14(11), 14895-14906 (2020-11-11)
The ability to apply force and torque directly to micro- and nanoscale particles in optical traps has a wide range of applications. While full control of both force and torque in three dimensions has been realized using top-down fabrication of
Seungkyu Ha et al.
ACS photonics, 6(5), 1255-1265 (2019-05-24)
Optical trapping of (sub)micron-sized particles is broadly employed in nanoscience and engineering. The materials commonly employed for these particles, however, have physical properties that limit the transfer of linear or angular momentum (or both). This reduces the magnitude of forces
Marcella Brescia et al.
International journal of molecular sciences, 21(19) (2020-10-01)
Duchenne muscular dystrophy (DMD) is the most frequent and severe form of muscular dystrophy. The disease presents with progressive body-wide muscle deterioration and, with recent advances in respiratory care, cardiac involvement is an important cause of morbidity and mortality. DMD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service