Skip to Content
MilliporeSigma
All Photos(1)

Documents

12019

Sigma-Aldrich

1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, 7-nitrobenzofurazan-labeled

for fluorescence, ammonium salt, ≥98.0% (TLC)

Synonym(s):

2(R),3-dipalmitoylglycero-1 -phospho-N-(7-nitro-2,1,3-benzoxadiazo1-4-y1)ethanolamine, L-β,γ-Dipalmitoyl-α-cephaline, 7-nitrobenzofurazan-labeled, N-(7-Nitro-benzofurazan-4-yl)-1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine, N-(7-Nitrobenzofurazan-4-yl)-1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine, N-NBD-Dipalmitoyl-L-α-Phosphatidylethanolamine, NBD-PE, Phosphatidylethanolamine 16:0, NBD-labeled

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H75N4O11P
CAS Number:
Molecular Weight:
855.05
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for fluorescence

Assay

≥98.0% (TLC)

fluorescence

λex 459 nm; λem 528 nm in methanol

storage temp.

−20°C

SMILES string

O=C(CCCCCCCCCCCCCCC)OC[C@@H](OC(CCCCCCCCCCCCCCC)=O)COP(OCCNC1=CC=C([N+]([O-])=O)C2=NON=C12)(O)=O

InChI

1S/C43H75N4O11P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-40(48)54-35-37(57-41(49)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)36-56-59(52,53)55-34-33-44-38-31-32-39(47(50)51)43-42(38)45-58-46-43/h31-32,37,44H,3-30,33-36H2,1-2H3,(H,52,53)/t37-/m1/s1

InChI key

ODWMPKRLTCWGFP-DIPNUNPCSA-N

Application

Phosphatidylethanolamine 16:0 can be utilized as a lipid fluorescent probe to study the morphological stability of lipid membranes (to surfactants and ethanol) by monitoring its fluorescence.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsuo Mioka et al.
Molecular biology of the cell, 29(10), 1203-1218 (2018-03-16)
Phospholipid flippase (type 4 P-type ATPase) plays a major role in the generation of phospholipid asymmetry in eukaryotic cell membranes. Loss of Lem3p-Dnf1/2p flippases leads to the exposure of phosphatidylserine (PS) and phosphatidylethanolamine (PE) on the cell surface in yeast
Deepti Dabral et al.
Cells, 8(4) (2019-04-17)
The fundamental molecular mechanism underlying the membrane merger steps of regulated exocytosis is highly conserved across cell types. Although involvement of Phospholipase A₂ (PLA₂) in regulated exocytosis has long been suggested, its function or that of its metabolites-a lyso-phospholipid and
Takuo Osawa et al.
Nature structural & molecular biology, 26(4), 281-288 (2019-03-27)
A key event in autophagy is autophagosome formation, whereby the newly synthesized isolation membrane (IM) expands to form a complete autophagosome using endomembrane-derived lipids. Atg2 physically links the edge of the expanding IM with the endoplasmic reticulum (ER), a role
Tianshu Li et al.
Nanomedicine : nanotechnology, biology, and medicine, 13(3), 1219-1227 (2016-12-15)
1,5-Dihexadecyl N,N-diglutamyl-lysyl-L-glutamate (GGLG) liposomes were previously developed to enhance drug delivery efficiency in tumor cells owing to its pH-responsive properties. Herein, we report the modification of GGLG liposomes by conjugating a Fab' fragment of an ErbB2 antibody to the terminus
Shen Wang et al.
Nature communications, 10(1), 69-69 (2019-01-10)
The transition of the Munc18-1/syntaxin-1 complex to the SNARE complex, a key step involved in exocytosis, is regulated by Munc13-1, SNAP-25 and synaptobrevin-2, but the underlying mechanism remains elusive. Here, we identify an interaction between Munc13-1 and the membrane-proximal linker

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service