MilliporeSigma
  • Atg2 mediates direct lipid transfer between membranes for autophagosome formation.
Atg2 mediates direct lipid transfer between membranes for autophagosome formation.
Nature structural & molecular biology (2019-03-27)
Takuo Osawa, Tetsuya Kotani, Tatsuya Kawaoka, Eri Hirata, Kuninori Suzuki, Hitoshi Nakatogawa, Yoshinori Ohsumi, Nobuo N Noda
ABSTRACT
A key event in autophagy is autophagosome formation, whereby the newly synthesized isolation membrane (IM) expands to form a complete autophagosome using endomembrane-derived lipids. Atg2 physically links the edge of the expanding IM with the endoplasmic reticulum (ER), a role that is essential for autophagosome formation. However, the molecular function of Atg2 during ER-IM contact remains unclear, as does the mechanism of lipid delivery to the IM. Here we show that the conserved amino-terminal region of Schizosaccharomyces pombe Atg2 includes a lipid-transfer-protein-like hydrophobic cavity that accommodates phospholipid acyl chains. Atg2 bridges highly curved liposomes, thereby facilitating efficient phospholipid transfer in vitro, a function that is inhibited by mutations that impair autophagosome formation in vivo. These results suggest that Atg2 acts as a lipid-transfer protein that supplies phospholipids for autophagosome formation.