Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06788

Supelco

2-(4-Hydroxyphenylazo)benzoic acid

matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure

Synonym(s):

2-(4′-Hydroxybenzeneazo)benzoic acid, 4′-Hydroxyazobenzene-2-carboxylic acid, HABA, HBABA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4N=NC6H4CO2H
CAS Number:
Molecular Weight:
242.23
Beilstein:
1842696
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.5% (HPLC)

form

powder

mp

204-208 °C (lit.)
204-208 °C

solubility

ethanol: 20 mg/mL, clear

cation traces

Ag: ≤1.0 mg/kg
Al: ≤1.0 mg/kg
B: ≤1.0 mg/kg
Ba: ≤1.0 mg/kg
Be: ≤1.0 mg/kg
Bi: ≤1.0 mg/kg
Ca: ≤1.0 mg/kg
Cd: ≤1.0 mg/kg
Co: ≤1.0 mg/kg
Cr: ≤1.0 mg/kg
Cu: ≤1.0 mg/kg
Fe: ≤1.0 mg/kg
Ge: ≤1.0 mg/kg
K: ≤5 mg/kg
Li: ≤1.0 mg/kg
Mg: ≤1.0 mg/kg
Mn: ≤1.0 mg/kg
Na: ≤5 mg/kg
Ni: ≤1.0 mg/kg
Pb: ≤1.0 mg/kg
Sn: ≤1.0 mg/kg
Sr: ≤1.0 mg/kg
Tl: ≤1.0 mg/kg
Zn: ≤1.0 mg/kg

absorption

≥2000 at 337 nm (mol. absorption)

SMILES string

Oc1ccc(cc1)\N=N\c2ccccc2C(O)=O

InChI

1S/C13H10N2O3/c16-10-7-5-9(6-8-10)14-15-12-4-2-1-3-11(12)13(17)18/h1-8,16H,(H,17,18)/b15-14+

InChI key

DWQOTEPNRWVUDA-CCEZHUSRSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noritaka Kato et al.
The journal of physical chemistry. B, 109(42), 19604-19612 (2006-07-21)
This study reports a homogeneous and competitive fluorescence quenching immunoassay based on gold nanoparticle/polyelectrolyte (Au(NP)/PE) coated latex particles prepared by the layer-by-layer (LbL) technique. First, the resonant energy transfer from a layer of fluorescent PEs to Au(NP) in LbL assembled
Joan-Antoni Farrera et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(7), 2277-2285 (2007-12-29)
The tautomeric equilibria of 2-(4'-hydroxyphenylazo)benzoic acid (HABA) and 2-(3',5'-dimethyl-4'-hydroxyphenylazo)benzoic acid (3',5'-dimethyl-HABA) have been studied by a combination of spectroscopic and computational methods. For neutral HABA in solvents of different polarity (toluene, chloroform, DMSO, DMF, butanol, and ethanol) the azo tautomer
M A Domin et al.
Rapid communications in mass spectrometry : RCM, 13(4), 222-226 (1999-03-31)
The ability to rapidly identify the taxonomic class of the wide variety of microorganisms involved in human and animal disease is becoming increasingly important, especially with the increasing development of resistance to the antibiotics which form the main defence against
Juha A E Määttä et al.
Chembiochem : a European journal of chemical biology, 9(7), 1124-1135 (2008-04-03)
Chicken avidin is a key component used in a wide variety of biotechnological applications. Here we present a circularly permuted avidin (cpAvd4-->3) that lacks the loop between beta-strands 3 and 4. Importantly, the deletion of the loop has a positive
Kai Qi et al.
Journal of the American Chemical Society, 126(21), 6599-6607 (2004-05-27)
Shell cross-linked nanoparticles (SCKs) presenting surface- and bioavailable biotin functional groups were synthesized via a mixed micelle methodology, whereby co-micellization of chain terminal biotinylated poly(acrylic acid)-b-poly(methyl acrylate) (PAA-b-PMA) and nonbiotinylated PAA-b-PMA were cross-linked in an intramicellar fashion within the shell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service