Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05706

Sigma-Aldrich

DL-allo-Isoleucine

≥99.0%

Synonym(s):

(2RS,3SR)-2-Amino-3-methylpentanoic acid, DL-Alloisoleucine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$237.00

$237.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$237.00

About This Item

Linear Formula:
CH3CH2CH(CH3)CH(NH2)COOH
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721795
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$237.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99.0%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

impurities

≤0.6% DL-isoleucine (GC)

color

white

mp

~285 °C (dec.)

application(s)

detection
peptide synthesis

SMILES string

CC[C@H](C)[C@@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t4-,5+/m0/s1

InChI key

AGPKZVBTJJNPAG-CRCLSJGQSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service