Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03695

Sigma-Aldrich

Ethylenediaminetetraacetic acid tetrasodium salt dihydrate

purum, ≥98.0% (KT)

Synonym(s):

EDTA tetrasodium salt, Edathamil, Tetrasodium ethylenediaminetetraacetate dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$57.50
1 KG
$162.00

$57.50


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$57.50
1 KG
$162.00

About This Item

Linear Formula:
(NaOOCCH2)2NCH2CH2N(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
416.20
Beilstein:
3861753
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
form:
powder or crystals
solubility:
H2O: 0.1 g/mL, clear

$57.50


Estimated to ship onApril 07, 2025Details


Request a Bulk Order

grade

purum

Quality Level

Assay

≥98.0% (KT)

form

powder or crystals

reaction suitability

reagent type: chelator

impurities

5-10% water

solubility

H2O: 0.1 g/mL, clear

SMILES string

O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.4Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;;2*1H2/q;4*+1;;/p-4

InChI key

KSYNLCYTMRMCGG-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application


  • Evaluation of ethylenediaminetetraacetic acid, tetrasodium salt dihydrate (EDTA)-Tween 20 treatment versus protease digestion of formalin-fixed tissue sections for detection of bovine respiratory syncytial virus antigen in infected ovine lung.: This study compares the effectiveness of EDTA-Tween 20 treatment against protease digestion for detecting bovine respiratory syncytial virus antigen in formalin-fixed tissue sections of infected ovine lungs. The findings indicate that the EDTA-Tween 20 method is a viable alternative, offering a more efficient approach to antigen detection in veterinary pathology (Meehan et al., 1989).

Biochem/physiol Actions

Ethylenediaminetetraacetic acid (EDTA) is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service