Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00467

Sigma-Aldrich

Clarifying reagent for dairy products

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

density

0.97 g/mL at 20 °C

Quality Level

storage temp.

2-8°C

Application

Clarifying reagent for dairy products can be used in many chemical and enzymatic colorimetric measurements for estimating the quality of milk and dairy products. Clarifying reagent for dairy products renders opaque milk and dairy product samples transparent, which allows direct photometric measurements of the reaction mixtures. Clarifying reagent for dairy products has been used to optimize, validate and compare three methods of determining lipase activity in skim and whole milk.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - open cup

Flash Point(C)

20 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Bendicho et al.
Journal of dairy science, 84(7), 1590-1596 (2001-07-27)
Three methods of determining lipase activity were optimized, validated, and compared using skim and whole milk. A chromogenic ester (p-nitrophenyl caprylate) was used in all to quantify the enzyme activity through the release of p-nitrophenol. It was measured colorimetrically (method
Gérard Humbert et al.
The Journal of dairy research, 73(4), 464-471 (2006-07-13)
Difficulties with the analysis of opaque milk and dairy products can be resolved by rendering the sample transparent. This allows direct photometric measurements of the reaction mixtures, and all sample pre-treatment steps become unnecessary. For this, several reagents have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service