Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CC4000

Sigma-Aldrich

Urokinase-type Plasminogen Activator

Synonym(s):

uPA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$478.00

$478.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$478.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

$478.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

Assay

>98% (SDS-PAGE)

form

solid

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... PLAU(5328)

General description

A thrombolytic agent that converts plasminogen to plasmin. A mixture of high molecular weight and low molecular weight urokinase.
Product Source: Prepared from urine of healthy individuals

Physical form

Lyophilized from 200 mM manitol, 100 mM NaCl, 50 mM Tris-HCl, pH 7.5 and 0.1 % PEG.

Storage and Stability

Reconstitute using 200-400 μL dH2O. Store at -20°C. Following reconstitution, aliquot and freeze (-70°C). This product is stable for 2 year as supplied. Stock solutions are stable for up to 6 months at -70°C.

Analysis Note

High molecular weight urokinase (54 kD): 40%.Low molecular weight urokinase (33 kD): 50%.Transitory forms of urokinase: 10%

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thrombolytic therapy: a comparison between urokinase and streptokinase. From a national cooperative study.
W R Bell
Seminars in thrombosis and hemostasis, 2(1), 1-13 (1975-07-01)
Assay methods and standard preparations for plasmin, plasminogen and urokinase in purified systems, 1967-1968.
Johnson, A J, et al.
Thrombosis et Diathesis Haemorrhagica, 21, 259-272 (1969)
Sean T Zuckerman et al.
Biomaterials, 30(23-24), 3825-3833 (2009-05-16)
Mass spectrometry is a powerful proteomic tool enabling researchers to survey the global proteome of a cell. This technique has only recently been employed to investigate cell-material interactions. We had previously identified material scarcity and limited adherent cells as challenges

Questions

1–2 of 2 Questions  
  1. Hello, I am interested in adding uPA to the medium of my cell culture of human pluripotent stem cell. Is product number CC4000 suitabl and ready to add to the medium, or does the protein needs activation before usage?

    1 answer
    1. This product does not require activation. Note that this material has not been tested for use in cell culture. Suitability would have to be determined by the end user.

      Helpful?

  2. Is CC4000, Urokinase-type Plasminogen Activator is in its active conformation and ready to be used in protease activity assays, or it needs to be activated before use?

    Is CC4000, Urokinase-type Plasminogen Activator is in its active conformation and ready to be used in protease activity assays, or it needs to be activated before use?

    1 answer
    1. This product is a combination of active (low molecular weight) and inactive (high molecular weight) urokinase. The composition of the CC4000 product is as follows:
      Low molecular weight urokinase (33 kD) 50%
      High molecular weight urokinase (54 kD) 40%
      Transient forms of urokinase 10%
      Activation prior to use is not required.
      uPA is produced as an inactive form called pro-uPA which is 54KDa. Pro-uPA is cleaved by proteases to produce a low molecular weight active form that is 33 kDa. This cleavage by plasmin occurs in two steps. The transitory form is the result of the first cleavage step. The second cleavage of this transient form results in the active 33 kDa uPA. The link below may provide additional helpful information:

      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC5969797/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service