Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-2880

Sigma-Aldrich

Sodium hexametaphosphate

SAJ first grade, 65.0-83.0% P2O5 basis

Synonym(s):

Calgon, Phosphate glass, water soluble, Polyphosphate sodium salt, Sodium polyphosphate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

65.0-83.0% P2O5 basis

form

solid

availability

available only in Japan

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[P](=O)([O-])([O-])O[P](=O)(O)O

InChI

1S/2Na.H4O7P2/c;;1-8(2,3)7-9(4,5)6/h;;(H2,1,2,3)(H2,4,5,6)/q2*+1;/p-2

InChI key

GYQBBRRVRKFJRG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Intended for the cytologic demonstration of Naphthol AS-D Chloroacetate Esterase in blood, bone marrow films or tissue touch preparations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hironori Taniguchi et al.
Microbial cell factories, 18(1), 75-75 (2019-04-27)
Thermostable enzymes have several advantages over their mesophilic counterparts for industrial applications. However, trade-offs such as thermal instability of enzyme substrates or co-factors exist. Nicotinamide adenine dinucleotide (NAD+) is an important co-factor in many enzyme-catalyzed oxidation-reduction reactions. This compound spontaneously

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service