Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-2800

Sigma-Aldrich

L-Glutamic acid monosodium salt monohydrate

SAJ special grade, ≥99.0%

Synonym(s):

L-2-Aminopentanedioic acid, MSG, Sodium L-glutamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH2CH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
187.13
Beilstein:
4164348
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Assay:
≥99.0%
grade:
SAJ special grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

232 °C (dec.) (lit.)

application(s)

peptide synthesis

storage temp.

15-25°C

SMILES string

O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O

InChI

1S/C5H9NO4.Na.H2O/c6-3(5(9)10)1-2-4(7)8;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t3-;;/m0../s1

InChI key

GJBHGUUFMNITCI-QTNFYWBSSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service