Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-1630

Sigma-Aldrich

Alginic acid sodium salt

CP, viscosity 500 cP 

Synonym(s):

Algin, Sodium alginate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300

grade

CP

form

solid

availability

available only in Japan

viscosity

500 cP

InChI

1S/C6H10O7.Na/c7-1-2(8)4(5(10)11)13-6(12)3(1)9;/h1-4,6-9,12H,(H,10,11);/q;+1/p-1/t1-,2-,3-,4?,6+;/m0./s1

InChI key

MSXHSNHNTORCAW-MPGIDXPLSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie Vanacker et al.
Fertility and sterility, 99(5), 1363-1368 (2013-02-05)
To evaluate the survival and growth potential of human preantral follicles isolated before and after cryopreservation. Pilot study. Gynecology research unit in a university hospital. Six women aged 27 to 32 years. Six ovarian biopsy samples were cut into two
Li-Hsin Han et al.
Biomaterials, 34(17), 4251-4258 (2013-03-16)
Macropores in tissue engineering scaffolds provide space for vascularization, cell-proliferation and cellular interactions, and is crucial for successful tissue regeneration. Modulating the size and density of macropores may promote desirable cellular processes at different stages of tissue development. Most current
M J Martín-Villena et al.
Carbohydrate polymers, 94(1), 1-11 (2013-04-03)
To develop more effective antifungal microparticulate therapeutic systems for the treatment of Candida vaginitis, microparticles containing nystatin were elaborated by emulsification/internal gelation method. Three types of microparticles were successfully prepared, alginate microparticles, chitosan and poloxamer 407 coated alginate microparticles. DSC
A A Chaturvedi et al.
The British journal of surgery, 100(7), 904-910 (2013-04-18)
Postoperative adhesion formation is a common consequence of abdominal surgery, and constitutes a major source of morbidity and mortality. This study evaluated an ultrapure alginate-based antiadhesive barrier gel. Experiments were performed in a rat model with caecal abrasion and peritoneal
Sven E F Borgos et al.
BMC systems biology, 7, 19-19 (2013-03-19)
Alginate is an industrially important polysaccharide, currently produced commercially by harvesting of marine brown sea-weeds. The polymer is also synthesized as an exo-polysaccharide by bacteria belonging to the genera Pseudomonas and Azotobacter, and these organisms may represent an alternative alginate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service