MilliporeSigma
  • Prevention of postsurgical adhesions using an ultrapure alginate-based gel.
Prevention of postsurgical adhesions using an ultrapure alginate-based gel.
The British journal of surgery (2013-04-18)
A A Chaturvedi, R M L M Lomme, T Hendriks, H van Goor
ABSTRACT
Postoperative adhesion formation is a common consequence of abdominal surgery, and constitutes a major source of morbidity and mortality. This study evaluated an ultrapure alginate-based antiadhesive barrier gel. Experiments were performed in a rat model with caecal abrasion and peritoneal side wall excision. The primary endpoint was the incidence of adhesions at 14 days after surgery. In experiment 1 (24 rats), animals treated with alginate gel were compared with controls that had no antiadhesive barrier. In experiment 2 (42 rats), alginate gel was compared with sodium hyaluronate carboxymethyl cellulose (HA/CMC) membrane and with no antiadhesive barrier. To check for any remote action of the gel, in experiment 3 (45 rats) application of alginate gel to the ipsilateral versus contralateral side of injury was compared with no antiadhesive barrier. In experiment 1, ultrapure alginate gel reduced the incidence of adhesions from eight of 12 in control animals to one in 12 (P = 0·009). Tissue healing assessed by histology was similar in both groups. In experiment 2, ultrapure alginate gel and HA/CMC membrane showed similar antiadhesive effectiveness, reducing the incidence of adhesions from ten of 14 rats in the control group to three of 14 (P = 0·021) and two of 14 (P = 0·006) respectively. In experiment 3, ultrapure alginate gel reduced the incidence of adhesions at the site of direct application (1 of 15) compared with controls (13 of 15; P = 0·001), but not if applied remotely (9 of 15; P = 0·214). Ultrapure alginate gel decreased the incidence of postoperative adhesion formation in this rat model.
Sigma-Aldrich
Alginic acid from brown algae, powder
Sigma-Aldrich
Sodium alginate
Sigma-Aldrich
Alginic acid sodium salt, powder
Sigma-Aldrich
Alginic acid calcium salt from brown algae
Sigma-Aldrich
Alginic acid sodium salt from brown algae, Medium viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, low viscosity
Sigma-Aldrich
Alginic acid sodium salt from brown algae, BioReagent, suitable for immobilization of micro-organisms