Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-1245

Sigma-Aldrich

Silver diethyldithiocarbamate

JIS special grade, ≥98.0%, storage temp.:2-8°C

Synonym(s):

DETC, Diethyldithiocarbamic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCSSAg
CAS Number:
Molecular Weight:
256.14
Beilstein:
3718150
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥98.0%
grade:
JIS special grade
form:
solid
Pricing and availability is not currently available.

grade

JIS special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

172-175 °C (lit.)

storage temp.

2-8°C

SMILES string

CCN(CC)C(=S)S[Ag]

InChI

1S/C5H11NS2.Ag/c1-3-6(4-2)5(7)8;/h3-4H2,1-2H3,(H,7,8);/q;+1/p-1

InChI key

NSVHDIYWJVLAGH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Also used for colorimetric determination of small quantities of arsenic
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

Biochem/physiol Actions

Inhibits induction of macrophage nitric oxide synthase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjoy K Maji et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 42(7), 989-996 (2007-06-15)
The efficiency of a locally available laterite soil in removing both arsenite and arsenate from aqueous medium by adsorption was evaluated. It was observed that in batch experiment conducted at 0.5 mg/L initial concentration of arsenic, laterite soil could remove
D Brune et al.
Swedish dental journal, 4(3), 125-128 (1980-01-01)
The reliability of the method for analysis for arsenic in dental cements according to the international standard ISO 2590 has been reinvestigated using nuclear tracer techniques and intercomparison of results obtained for the arsenic content of various cements by means
Seth H Frisbie et al.
Environmental health perspectives, 113(9), 1196-1204 (2005-09-06)
All of Bangladesh's approximately 10 million drinking-water tube wells must be periodically tested for arsenic. The magnitude of this task and the limited resources of Bangladesh have led to the use of low-cost, semiquantitative field kits that measure As to
Yabing Xiao et al.
Water environment research : a research publication of the Water Environment Federation, 79(9), 1015-1022 (2007-10-04)
A novel method of preconcentration of trace arsenite and arsenate by using titanium dioxide nanoparticles as adsorbent was described. The concentrations of preconcentrated arsenite and arsenate were determined by a silver diethyldithiocarbamate spectrophotometric method without desorption. Batch adsorption experiments were
Mohammad Hossein Arbab-Zavar et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 26(1), 107-110 (2010-01-13)
A simple method was developed for speciation and spectrophotometric determination of inorganic As(III) and As(V) using an electrochemical hydride generation technique. For speciation of As(III) and As(V), a graphite rod was used as cathode to reduce As(III) to AsH(3), the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service