Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.93219

Supelco

Sodium citrate, dihydrate

for analysis EMPARTA® ACS

Synonym(s):

Sodium citrate tribasic dihydrate, Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COONa)(CH2COONa)2 · 2H2O
CAS Number:
Molecular Weight:
294.10
Beilstein:
6104939
MDL number:
UNSPSC Code:
12161703

grade

ACS reagent

mp

>300 °C (lit.)

storage temp.

room temp

SMILES string

O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.3Na.2H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;2*1H2/q;3*+1;;/p-3

InChI key

NLJMYIDDQXHKNR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Packaging

1.93219.0521: 500 g, Minimum order quantity: 1
1.93219.5021: 5 kg, Minimum order quantity: 1

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service