Skip to Content
MilliporeSigma
All Photos(1)

Documents

07-3000

Sigma-Aldrich

Diethylene glycol methyl ether

SAJ first grade, ≥99.0%

Synonym(s):

2-(2-Methoxyethoxy)ethanol, Methyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
120.15
Beilstein:
1697812
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.14 (vs air)

vapor pressure

0.2 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

221 °C

expl. lim.

1.38-22.7 %

availability

available only in Japan

refractive index

n20/D 1.426 (lit.)

bp

194 °C (lit.)

mp

−70 °C (lit.)

density

1.023 g/mL at 25 °C (lit.)

SMILES string

COCCOCCO

InChI

1S/C5H12O3/c1-7-4-5-8-3-2-6/h6H,2-5H2,1H3

InChI key

SBASXUCJHJRPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Zhu et al.
Chemical communications (Cambridge, England), 48(96), 11766-11768 (2012-11-02)
Rlyso, a highly selective and sensitive pH sensor, can stain lysosomes with a novel lysosome-locating group, methylcarbitol. Rlyso was successfully used to detect lysosomal pH changes during apoptosis or induced by chloroquine while avoiding the "alkalizing effect" on lysosomes of
R R Miller et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 5(6 Pt 1), 1174-1179 (1985-12-01)
Male and female Fischer 344 rats were exposed to 0, 30, 100, or 216 ppm diethylene glycol monomethyl ether (DEGME) vapors (0, 0.15, 0.49, or 1.06 mg/liter) 6 hr/day, 5 days/week, for 13 weeks. The 216-ppm exposure level was the
B D Hardin et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 6(3), 430-439 (1986-04-01)
Diethylene glycol monomethyl ether (diEGME) was one of 15 glycols tested in CD-1 mice using a short-term in vivo reproductive toxicity assay (Chernoff/Kavlock test). Because results were strongly suggestive of potential reproductive toxicity, a teratology study was conducted in Sprague-Dawley
T Kawamoto et al.
Toxicology, 76(1), 49-57 (1992-11-22)
In this paper, we determined whether ethylene glycol monomethyl ether (EGME) and diethylene glycol monomethyl ether (diEGME) induce hepatic gamma-glutamyl transpeptidase activity. Male adult Wistar rats weighing 220 g were used as experimental animals. EGME (100, 300 mg/kg per day)
M Ihsan Karaman et al.
Journal of pediatric surgery, 37(8), E23-E23 (2002-08-01)
Retrocaval ureter is a very rare condition. In light of the experimental studies, one of the etiologic factors seems to be maternal contact with diethylene glycol monomethyl ether or ethylene glycol monomethyl ether. A case of cardiovascular, skeletal, and retrocaval

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service