Skip to Content
MilliporeSigma
All Photos(1)

Documents

266558

Sigma-Aldrich

Aluminum

wire, diam. 1.0 mm, 99.999% trace metals basis

Synonym(s):

Aluminium wire

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
EC Number:
MDL number:
UNSPSC Code:
12141702
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

wire

autoignition temp.

1400 °F

resistivity

2.6548 μΩ-cm

diam.

1.0 mm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

Related Categories

General description

Aluminum (Al) is a lightweight, malleable metal known for its excellent corrosion resistance and electrical conductivity. This product is available in wire form with a diameter of 1.0 mm, featuring a resistivity of 2.6548 μΩ-cm. With an assay of 99.999% trace metals basis, aluminum has a melting point of 660.37 °C and a density of 2.7 g/mL at 25 °C. Aluminum is extensively utilized in various applications, including aerospace and automotive, due to its strength-to-weight ratio and durability. It is commonly used in the production of lightweight components, structural materials, and packaging solutions. Additionally, aluminum′s high conductivity makes it an ideal choice for electrical wiring and components.

Application

  • A lithium-aluminium heterobimetallic dimetallocene.: This paper introduces a novel bimetallic compound combining lithium and aluminium, offering insights into their cooperative effects in chemical synthesis and potential applications in material science (Bischoff IA et al., 2024).
  • Tridentate Lewis Acids Based on Tribenzotriquinacene Chalices.: Discusses the synthesis of new tridentate Lewis acids using tribenzotriquinacene frameworks, enhancing our understanding of their chemical behavior and applications in catalysis (Franke M et al., 2024).

Quantity

  • 5 m (approximately 10.5 g)
  • 25 m (approximately 52.5 g)

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thierry Deschamps et al.
Optics express, 21(7), 8382-8392 (2013-04-11)
Using a combination of experimental techniques such as optical absorption, Raman scattering, continuous wave and pulse Electron Spin Resonance (ESR), we characterize a set of γ-irradiated Yb(3+) doped silica glass preforms with different contents of phosphorous and aluminum. We demonstrate
Caitlin Carney et al.
Journal of the American Academy of Dermatology, 69(4), 578-582 (2013-07-17)
Laser treatment has emerged as a novel treatment modality for onychomycosis. We sought to determine thermal response and optical effects of a submillisecond neodymium:yttrium-aluminum-garnet (Nd:YAG) 1064-nm laser on common fungal nail pathogens, and the clinical efficacy and safety of the
Arjun Joshua et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(24), 9633-9638 (2013-05-28)
Controlling the coupling between localized spins and itinerant electrons can lead to exotic magnetic states. A novel system featuring local magnetic moments and extended 2D electrons is the interface between LaAlO3 and SrTiO3. The magnetism of the interface, however, was
Jesper C Holst et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), 8819-8823 (2013-05-15)
Refractory inclusions [calcium-aluminum-rich inclusions, (CAIs)] represent the oldest Solar System solids and provide information regarding the formation of the Sun and its protoplanetary disk. CAIs contain evidence of now extinct short-lived radioisotopes (e.g., (26)Al, (41)Ca, and (182)Hf) synthesized in one
Julian I Schroeder et al.
Nature, 497(7447), 60-66 (2013-05-03)
With the global population predicted to grow by at least 25 per cent by 2050, the need for sustainable production of nutritious foods is critical for human and environmental health. Recent advances show that specialized plant membrane transporters can be

Articles

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service