Skip to Content
MilliporeSigma
All Photos(1)

Documents

T4PNK-RO

Roche

T4 Polynucleotide Kinase, 3′-phosphatase free

from phage T4 am N81 pse T1 infected Escherichia coli BB

Synonym(s):

kinase, t4 polynucleotide, 3′-phosphatase-free

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

solution

specific activity

≥40 x 103 units/mg protein
40000 U/mg

packaging

pkg of 1,000 U (10838292001)
pkg of 200 U (10709557001)

manufacturer/tradename

Roche

storage temp.

−20°C

General description

T4 polynucleotide kinase catalyzes the transfer of the terminal phosphate group of ATP to the 5′-hydroxyl terminus of DNA or RNA. It also can catalyze the exchange of 5′-terminal phosphate groups. In contrast to the wild type E. coli enzyme, this phage derivative lacks 3′-phosphatase activity. The assay time is 30 minutes and volume activity is 10000 U/ml.

Application

Use T4 Polynucleotide Kinase, 3′-phosphatase-free, to:
  • Phosphorylate the 5′ ends of RNA or DNA
  • Phosphorylate the 3′ ends of RNA to prevent cyclization
  • Add a 5′[32P]-terminal label to 3′-CMP, forming 5′[32P]pCp. This substrate is commonly used for 3′-end labeling of RNA with T4 RNA ligase.
  • Label 5′-hydroxyl ends of DNA with [?-32P]-ATP (either by direct phosphorylation or by phosphate exchange)
  • Phosphorylate the 5′-hydroxyl ends of oligonucleotides obtained from automated synthesizers
  • Phosphorylate the 5′-hydroxyl ends of linkers.
  • Purify recombinant RNA in E. coli.

Note: The 3′-phosphatase activity of the wild-type kinase is not observed under optimal incubation conditions with the mutant T4 Polynucleotide Kinase.

Biochem/physiol Actions

T4 polynucleotide kinase catalyzes the transfer of the terminal phosphate group of ATP to the 5′-hydroxyl terminus of DNA or RNA. It also can catalyze the exchange of 5′-terminal phosphate groups. In contrast to the wild type E. coli enzyme, this phage derivative lacks 3′-phosphatase activity. The assay time is 30 minutes and volume activity is 10000 U/ml.

Packaging

1 kit containing 2 components

Quality

Absence of 3′-phosphatase, endonucleases, nicking activity, 3′-exonucleases, and ribonucleases, tested according to the current Quality Control procedures; function tested using the DNA 5′-End Labeling Kit.

Unit Definition

One unit is the enzyme activity which catalyzes the incorporation of 1 nmol 32P into acid-precipitable products within 30 minutes at +37 °C.

Volume Activity: 10 x 103 U/ml

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Enzyme Solution, in buffer, pH 7.5 (+25 °C) 10 U/μl

  • Phosphorylation Buffer 10x concentrated

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorylating DNA with DNA
Li Y and Breaker RR
Proceedings of the National Academy of Sciences of the USA, 2746-51 null
Luc Ponchon et al.
Nature protocols, 4(6), 947-959 (2009-05-30)
RNA production using in vivo transcription by Escherichia coli allows preparation of milligram quantities of RNA for biochemical, biophysical and structural investigations. We describe here a generic protocol for the overproduction and purification of recombinant RNA using liquid chromatography. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service