Skip to Content
MilliporeSigma
All Photos(1)

Documents

11699113001

Roche

MgCl2 Stock Solution

pH 8.25-8.45 (25 °C), suitable for PCR, solution, pkg of 3 × 1 mL

Synonym(s):

magnesium chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

form

solution

Quality Level

packaging

pkg of 3 × 1 mL

manufacturer/tradename

Roche

concentration

25 mM

technique(s)

PCR: suitable

pH

8.25-8.45 (25 °C)

storage temp.

−20°C

General description

The MgCl2 Stock Solution is used for the individual adjustment of the Mg2+-concentration in the PCR reaction and can be used in combination with the PCR buffer without MgCl2. In most applications a concentration of 1.5mM MgCl2 will yield satisfactory results with a dNTP concentration of 200μM each. In many cases, however, it is required to titrate the optimal Mg2+ concentration to increase the specificity and the yield of the PCR reaction.

Application

The combination of PCR Buffer without MgCl2 and the MgCl2 Stock Solution buffer are optimized for the amplification of specific DNA fragments in conventional PCRs (i.e., with Taq DNA Polymerase). However, the MgCl2 Stock Solution may be used with other, more appropriate PCR buffers to optimize any type of PCR (e.g., hot start PCR, high fidelity PCR, long template PCR).

Biochem/physiol Actions

Magnesium (Mg2+) chelates nucleotide triphosphates (dNTPs) and its usage is one among the optimization strategies for polymerase chain reaction (PCR). It promotes orthophosphate synthesis from oligophosphates by stabilizing the phosphate groups of nucleotides.

Physical form

The MgCl2 Stock Solution contains 25mM MgCl2 in sterile water.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The significance of Mg in prebiotic geochemistry
Holm NG
Geobiology, 10(4), 269-279 (2012)
Optimization and troubleshooting in PCR
Roux KH
Cold Spring Harbor Protocols, 2009(4), pdb-ip66 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service