Key Documents
WP61RBD00
Millipore
Chemical Duty Pump rebuild kit
Chemical Duty Pump rebuild kit
1 of 1 reviewers received a sample product or took part in a promotion
Select a Size
1 KIT
$505.00
$505.00
Please contact Customer Service for Availability
Select a Size
Change View
1 KIT
$505.00
About This Item
UNSPSC Code:
41105101
eCl@ss:
32012303
NACRES:
NB.24
$505.00
Please contact Customer Service for Availability
shipped in
ambient
General description
This item includes parts for catalog numbers WP6111560, WP6122050, WP6110060
This item is a kit composed of the following items:
(1) Diaphragm hold-down plate assembly, (1) Valve plate assembly, (1) Pump head, (1) Vacuum gauge, (1) Pressure gauge, (4) Head screws, (2) Handle screws
Reviews
1 of 1 reviewers received a sample product or took part in a promotion
Active Filters
  1. IL
    • Review 1
    • Votes 2
    5 out of 5 stars.
    Broken pump fixed
    Easy to replace, pump was fixed with no issues after. I wish, there were flapper valves sold separately, without the need to buy expensive kit.
    Helpful?