Skip to Content
MilliporeSigma
All Photos(2)

Documents

W430-450

Phosphate Buffer

vial of 450 mL, pkg of 20 units, bag W 6.5 in. , with a linear tear and heat induction seal, USP Butterfield′s Method, plastic bag (aseptically filled, sterile), sterile; γ-irradiated by SER-TAIN process

Synonym(s):

Phosphate Buffer, Remel Phosphate Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

material

plastic bag (aseptically filled, sterile)

Quality Level

Agency

USP Butterfield′s Method

sterility

sterile; γ-irradiated by SER-TAIN process

form

liquid

packaging

pkg of 20 units
vial of 450 mL

technique(s)

microbiological culture: suitable

bag W

6.5 in. , with a linear tear and heat induction seal

pH

7.2±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phosphate buffer is a liquid medium designed for the qualitative detection of microorganisms in pharmaceutical and food samples. It serves as a sample diluent and as a medium for performing serial dilution. It prevents pH-associated variations while carrying out microbial experiments.

Application

Phosphate buffer is used for sample dilutions to detect microorganisms in pharmaceutical and food samples.

Legal Information

SER-TAIN is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service