Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V0CPAK001

Milli-Q

VOC-Pak® Polisher

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$647.00

$647.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
$647.00

About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.14

$647.00


Please contact Customer Service for Availability

Request more information

material

carbon (activated)
polyethylene (Luer Plug)
polyethylene connector
polypropylene housing

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

VOC-Pak®

parameter

≤2 bar pressure
≤2.0 L/min flow rate
5 °C temp. range (30 °C)

resistivity

>18.0 MΩ-cm (at 25 degree celcius)

L

12.7 cm

impurities

<5 ppb TOC

fitting

1/4 in. inlet connection ( Gaz (straight) F connection and Luer F)
1/4 in. outlet connection (Gaz (straight) F connection and Luer F)

compatibility

for use with Direct-Q® UV
for use with Direct-Q®
for use with Milli-Q® Direct
for use with Milli-Q® Reference
for use with Simplicity® UV
for use with Simplicity®
for use with Synergy® UV
for use with Synergy®

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Production of volatile organic compound-free ultrapure water at the point of dispense of the Direct-Q® / Simplicity® / Synergy® / Milli-Q® Direct and Reference systems
VOC-Pak® Polisher is designed to purify ultrapure water in the final purification step at point-of-dispense of Type 1 water purification systems. It is an activated carbon-based cartridge specifically designed for the effective removal of volatile organic compounds (VOCs) from ultrapure water.

VOCs are environmental contaminants causing health and safety issues that needs to be routinely analyzed in water, soil, gas and air samples. Specifications for VOCs in drinking water have been established by different regulatory agencies in many countries.
VOC-Pak® Polisher is, therefore, an excellent choice for scientists in need of VOC-free ultrapure water for their analytical applications.

Application

The VOC-Pak Polisher is used to produce VOC-free water for the analysis of VOCs in water by GC and GC-MS when fed by an ultrapure water system.

VOC-free water is suitable for glassware rinsing, blanks, reagents, sample preparation, standard solutions or dilutions in the analysis of those substances, or in studies where VOCs may interfere. It is used in the development of nanoparticle sensors[1][2] and nanoelectrodes.

Features and Benefits

  • Validated to produce 300 L of VOC-free ultrapure water.
  • Easy maintenance: the polisher is easily installed and replaced.
  • Bottom-tip is redesigned for a secured installation of the protection bell.
  • e-Sure tag for RFID connection with Q-POD® dispenser enables full traceability (data management) and automatic consumable status monitoring on the POD′s touchscreen interface.
  • Certificate of Quality available online.
  • Provides a warranty of water suitable for VOC analysis: blanks and standards preparation; glassware cleaning; and sample dissolution (if VOCs are analyzed in a matrix other than water i.e., in soil).

Legal Information

DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany
SIMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
SYNERGY is a registered trademark of Merck KGaA, Darmstadt, Germany
VOC-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemistry at a single nanoparticle: from bipolar regime to tunnelling
Sun, T. et al.
Faraday Discussions, 210, 173-188 (2018)
J S Cooper et al.
The Analyst, 140(9), 3233-3238 (2015-03-15)
Partially selective gold nanoparticle sensors have the sensitivity and selectivity to discriminate and quantify benzene, toluene, ethylbenzene, p-xylene and naphthalene (BTEXN) at concentrations relevant to the US Environmental Protection Agency. In this paper we demonstrate that gold nanoparticle chemiresistors can

Questions

1–10 of 10 Questions  
  1. What are the storage conditions for this product?

    1 answer
    1. Recommended storage conditions: temperatures from 5 to 30°C; humidity between 10 and 80%. The original sealed bag (if provided) must be kept intact before the first use to protect the purification element from foreign matter.

      Helpful?

  2. How are consumables changed?

    1 answer
    1. To help you to change your system's consumables, several video tutorials are available on our YouTube channel: https://youtube.com/playlist?list=PL2TUriaAaEA_IXDCnPbDk6OmrwptjwOHD. For high-flow systems, please keep in mind that all consumable changes must be approved by a 3-digit code that only the person in charge of the system knows.

      Helpful?

  3. How can I know which consumables can be used with my system?

    1 answer
    1. To know which consumables can be used on your system, you can see in the user manual of your system or see all the consumables for each system listed on https://www.sigmaaldrich.com/products/water-purification/milli-q-consumables.

      Helpful?

  4. What steps are you taking towards improved sustainability?

    1 answer
    1. Sustainability is at the heart of our innovations. From reducing water and energy consumption, to minimizing plastic and chemical waste, our dedicated water system engineers are constantly working on new ways to reduce the ecological footprint of our products to better support the sustainability goals we're all striving towards. For more information, we invite you to review the following brochure and infographic which highlight our efforts and achievements: https://www.sigmaaldrich.com/life-science/milliq#sus. Do not hesitate to contact your local representative or sales contact (https://www.sigmaaldrich.com/collections/offices) for further information.

      Helpful?

  5. Can I arrange to have my consumables automatically ordered and delivered once a year?

    1 answer
    1. It's possible to place only one consumable order per year with a specific contract called "Consumable Supply Agreement (CSA)" (https://www.sigmaaldrich.com/US/en/products/water-purification/milli-q-consumables#csa). For further information, contact your local representative or sales contact (https://www.sigmaaldrich.com/US/en/collections/offices).

      Helpful?

  6. What is the consumables delivery time?

    1 answer
    1. Refer to the consumable's Product page for a first estimation of its expected delivery time and contact customer service for more information.

      Helpful?

  7. How frequently do the various consumables need to be changed?

    1 answer
    1. The frequency that consumables must be changed depends on the system's usage (treated water volume) and on the consumables themselves (time of usage). In order to ensure the quality of water and your results, the consumables should be replaced when the system alerts you.

      Helpful?

  8. If I have a final filter that I use only occasionally (e.g., Biopak®) and have another filter (e.g. Millipak®) that I most of the time, can I keep the 'occasional-use' filter for use again at a later time?

    1 answer
    1. It is not possible to re-use a filter that has been used and then removed because air intake during replacement creates an airlock and blocks water passing through the filter. If you regularly need two different filters, we recommend using two POD dispensers.

      Helpful?

  9. Where can I find data sheets, Certificates of Origin, Certificates of Analysis, and Certificates of Quality?

    1 answer
    1. Documents and certificates are available in the "Documentation" section on this page. Do not hesitate to contact us if you require further information.

      Helpful?

  10. Can consumables be opened and refilled?

    1 answer
    1. Unfortunately, this is not possible because of safety issues and the need to guarantee the quality of consumables and their interior purification components. Our consumables are manufactured in an ISO 9001-certified facility France in agreement with high quality standards.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service