Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EDSKIT001

Milli-Q

Application-Pak Installation Kit

For use with various lab water purification systems

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$272.00

$272.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EA
$272.00

About This Item

UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
NA.21

$272.00


Please contact Customer Service for Availability

Request more information

packaging

pkg of 1 kit

L

12.7 cm

compatibility

for use with EDS-Pak®
for use with LC-Pak®

shipped in

ambient

General description

Cartridge installation and Conditioning Kit includes the following reusable parts:
  • Polyethylene ¼ Gaz F-Hose barb connection with O-ring
  • Polyethylene ¼ G F – ¼ Gaz F connectors for connection to MILLIPAK®
  • A protection bell

Application

For use with LC-Pak®/EDS-Pak® Polisher. This kit is used for the initial installation and activation of Application paks.

Legal Information

EDS-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
LC-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIPAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service