Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

USBA0100LC1S1

Millipore

Mobius®, Silver, Ultimus® Bin Assembly

Collapsible bin, bottom-ported, 100L

Synonym(s):

Mobius®, Silver, Ultimus® Bin Assembly

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123312

material

(Ultimus® Film)
C-Flex
polypropylene
silicone

Quality Level

Agency

meets EN 2.6.14
meets ISO 10993-4
meets JP 4.01
meets USP 661
meets USP 788
meets USP 85
meets USP 87
meets USP 88

sterility

sterile; γ-irradiated (at 25-40 kGy)

product line

Mobius®

parameter

2—60 °C temp. range (35.6—140 °F)

capacity

100 L

impurities

<0.25 EU/mL Aqueous extraction (LAL test)

application(s)

pharma/biopharma processes

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Ultimus® film was designed for more challenging single-use applications such as large-volume liquid processing. Our Ultimus® film technology provides enhanced bag strength, improved durability and leak resistance through a novel strength layer reinforced by woven nylon.
Ultimus® Film now empowers Mobius® MIX and
Mobius® Power MIX Single-Use assemblies of 100L, 200L, 250L, 500L, 1000L and 2000L and is also available in our Mobius® 3D
assemblies and Mobius® iFlex Bioreactor assemblies.

Application

Buffer, media, product transportation, Intermediate fluid transfer and storage

Features and Benefits

Ultimus® Film technology provides enhanced bag strength, improved durability and leak resistance.

Packaging

Pack of 1 assembly

Components

Bag: Ultimus® Film, Polyolefin
Tubing: TPE, Platinum-cured sillicone.
Coupler: PP
Luer: PP
Adapter SterlEnz: PP, Silicone
Plug end: PP.
Pinch clamp: Polyester, PBT

Legal Information

MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTIMUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Shipping and storage of valuable bioprocessing liquids deserves the superior strength of single-use assemblies made with Ultimus® film.

Related Content

Film selection is guided by process needs, compatibility of the process fluid with film, process volume and risk. This brief compares the properties of our films so you can make the best selection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service