Skip to Content
MilliporeSigma
All Photos(3)

Documents

TZA000010

Millipore

Steridilutor® NEO Device

For liquid transfer from ampoules or vials to a diluent vial with septum pooling.

Synonym(s):

Steridilutor® NEO device for sterility testing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.24

material

Nylon 66 adapter (for needle)
PVC tubing
stainless steel (for needle)

Quality Level

Agency

EP (2.6.1)
JP (4.06)
USP 71

sterility

sterile; γ-irradiated

manufacturer/tradename

Steridilutor®

packaging

pkg of 10 blisters per box, Single packed

input

sample type pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)

shipped in

ambient

General description

The Steridilutor® NEO device for liquid transfer kit enables to dilute liquids from open ampoules into a diluent container with septum. For liquid antibiotics, a dilution step is strongly recommended to reduce the concentration of the antimicrobial agent. This will reduce the possibility of binding of the agent to the filter. The liquid transfer kit single needle is a tubing and needle assembly without vent. Small diameter double needle connects test product to diluent. The assembly is to be used for difficult to dissolve powders, dilution and pooling of viscous products in vials as well as antibiotics. The diluted products are subsequently tested with suitable Steritest® Neo canister.

Application

Steridilutor® NEO devices for liquid transfer is used for the transfer of liquids from ampoules or vials to a diluent vial with septum pooling. The tubing and needle assembly is also used to dilute and pool products in vials.

Legal Information

STERIDILUTOR is a registered trademark of Merck KGaA, Darmstadt, Germany
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service