Skip to Content
MilliporeSigma
All Photos(3)

Documents

SYMBFHA01

Millipore

Steritest® Symbio FLEX handles

Optional accessory, for use with Steritest® Symbio pump

Synonym(s):

Pump carrying aid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

material

316L stainless steel

Agency

EP (2.6.1)
JP (4.06)
USP (71)

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® Symbio FLEX handles are optional accessories on the FLEX version of the Steritest® Symbio pump. They can be added to your Steritest® Symbio FLEX pump when used on feet in a vertical laminar flow hood or isolator without a cutout.

Application

Steritest® Symbio FLEX handles are used to ease the transportation of the pump.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service